Skip to Content
Merck
All Photos(1)

Key Documents

175889

Sigma-Aldrich

tert-Butanol-d10

99 atom % D

Synonym(s):

2-Methyl-2-propanol-d10, tert-Butyl alcohol-d10

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€169.00
5 G
€582.00

€169.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€169.00
5 G
€582.00

About This Item

Linear Formula:
(CD3)3COD
CAS Number:
Molecular Weight:
84.18
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

€169.00


Please contact Customer Service for Availability

Request a Bulk Order

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

impurities

≤0.5% water
water

refractive index

n20/D 1.3835 (lit.)

bp

82 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

mass shift

M+10

SMILES string

[2H]OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3,3D3,5D

InChI key

DKGAVHZHDRPRBM-SGLLWXCUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butanol-d10 (tert-Butyl alcohol-d10) is a deuterated derivative of tert-butanol. It participates in the synthesis of cyanoacetic acid-tert-butylester-d9.[1] Conformational analysis of 2,3-dihydroxypropanoic acid in tert-butyl alcohol-d10 indicates the dominance of gauche-hydroxyl rotamers (92%) under low pH conditions.[2]

Application

tert-Butanol-d10 may be used in the synthesis of cyanoacetic acid-tert-butylester-d9[1] and tert-butoxide-d9 (BuOK-d9).[3]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silacyclobutane as ?Carbanion Pump? in Anionic Polymerization. III. Synthesis of Di-and Tri-block Copolymer by ?Diphenylsilacyclobutane-Potassium tert-Butoxide System
Hyun J-Y and Kawakami Y
Polymer Journal, 36(10), 856-865 (2004)
Thorsten W Jaskolla et al.
Journal of the American Society for Mass Spectrometry, 22(6), 976-988 (2011-09-29)
This work experimentally verifies and proves the two long since postulated matrix-assisted laser desorption/ionization (MALDI) analyte protonation pathways known as the Lucky Survivor and the gas phase protonation model. Experimental differentiation between the predicted mechanisms becomes possible by the use
Michael D Drake et al.
Magnetic resonance in chemistry : MRC, 44(3), 210-219 (2006-02-16)
Vicinal (1)H--(1)H coupling constants were used to determine the conformational preferences of 2,3-dihydroxypropanoic acid (1) (DL-glyceric acid) in various solvents and its different carboxyl ionization states. The stereospecific assignments of J(12) and J(13) were confirmed through the point-group substitution of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service