Skip to Content
Merck
All Photos(1)

Key Documents

168343

Sigma-Aldrich

N,N-Diethyl-p-phenylenediamine sulfate salt

97%

Synonym(s):

4-Amino-N,N-diethylaniline sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4NH2 · H2SO4
CAS Number:
Molecular Weight:
262.33
Beilstein:
4219768
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

184-186 °C (lit.)

SMILES string

OS(O)(=O)=O.CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2.H2O4S/c1-3-12(4-2)10-7-5-9(11)6-8-10;1-5(2,3)4/h5-8H,3-4,11H2,1-2H3;(H2,1,2,3,4)

InChI key

AYLDJQABCMPYEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne Paquin et al.
Arzneimittel-Forschung, 55(7), 359-369 (2005-08-06)
Oxidative stress may involve overproduction of hydrogen peroxide which can generate highly cytotoxic hydroxyl radicals in the presence of ferrous ions. This work demonstrates that TCAT (Tricomponent Antioxidant Therapy), an association of pyruvate, vitamin E and fatty acids, provides neuronal
Measurement of the effect of a plant-based diet on reactive oxygen metabolite production using the d-Roms test is invalid.
M I Harma et al.
The International journal of biological markers, 21(1), 60-60 (2006-05-23)
Anita Rakić et al.
Annals of agricultural and environmental medicine : AAEM, 19(3), 431-436 (2012-10-02)
The microbiological colonisation of buildings and man-made structures often occurs on the walls of plumbing systems; therefore, monitoring of opportunistic pathogens such as Legionella pneumophila (L. pneumophila), both in water distribution mains and in consumers' plumbing systems, is an important
Xiao-jie Xu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(10), 1235-1237 (2005-03-12)
In this paper, the absorption spectra of free chlorine in the visible region has been measured after free chlorine in the solution had reacted with N,N-diethyl-1,4-phenylenediamine (DPD). The relation between the absorption spectra and peak wavelength has been discussed. From
Z B Ogel et al.
Applied microbiology and biotechnology, 71(6), 853-862 (2006-01-04)
Scytalidium thermophilum produces an extracellular phenol oxidase on glucose-containing medium. Certain phenolic acids, specifically gallic acid and tannic acid, induce the expression of the enzyme. Production at 45 degrees C in batch cultures is growth-associated and is enhanced in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service