Skip to Content
Merck
All Photos(1)

Key Documents

167576

Sigma-Aldrich

2-Chlorobenzothiazole

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€45.80
25 G
€137.00

€45.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€45.80
25 G
€137.00

About This Item

Empirical Formula (Hill Notation):
C7H4ClNS
CAS Number:
Molecular Weight:
169.63
Beilstein:
116316
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€45.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.637 (lit.)

bp

141 °C/30 mmHg (lit.)

mp

21-23 °C (lit.)

density

1.303 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1nc2ccccc2s1

InChI

1S/C7H4ClNS/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H

InChI key

BSQLQMLFTHJVKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chlorobenzothiazole was used in the synthesis of:
  • (RS)- and (S)-lubeluzole[1]
  • (1,3-benzothiazol-2-yl) amino-9-(10H)-acridinone derivatives[2]
  • 4H-thieno[2′,3′:4,5]pyrimido[2,1-b]benzothiazole derivatives[3]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new thienopyrimidobenzothiazoles and thienopyrimidobenzoxazoles with analgesic and antiinflammatory properties.
Russo F, et al.
European Journal of Medicinal Chemistry, 29(7), 569-578 (1994)
Florence Delmas et al.
European journal of medicinal chemistry, 39(8), 685-690 (2004-07-28)
(1,3-Benzothiazol-2-yl) amino-9-(10H)-acridinone derivatives were synthesized via a procedure based on the Ullman reaction and were assessed for their in vitro antileishmanial and anti-HIV activities. Two derivatives, 4-(6-nitro-benzothiazol-2-ylamino)-10H-acridin-9-one and 1-(6-amino-benzothiazol-2-ylamino)-10H-acridin-9-one, revealed a selective antileishmanial activity, mainly due to amastigote-specific toxicity. Results
Damodara N Kommi et al.
Organic letters, 15(6), 1158-1161 (2013-02-26)
Three new, concise, and protecting group-free synthetic routes for (RS)- and (S)-lubeluzole are reported in higher (46-62%) overall yields compared to the reported procedures (6-35%). The key steps involve C-N bond formation via epoxide aminolysis and nucleophilic substitution of 2-chlorobenzothiazole
Anita K Kovács et al.
Frontiers in chemistry, 6, 120-120 (2018-05-05)
A general strategy for the synthesis of N-peptide-6-amino-D-luciferin conjugates has been developed. The applicability of the strategy was demonstrated with the preparation of a known substrate, N-Z-Asp-Glu-Val-Asp-6-amino-D-luciferin (N-Z-DEVD-aLuc). N-Z-DEVD-aLuc was obtained via a hybrid liquid/solid phase synthesis method, in which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service