Skip to Content
Merck
All Photos(1)

Key Documents

167037

Sigma-Aldrich

3-Methylsalicylic acid

96.0%

Synonym(s):

2-Hydroxy-3-methylbenzoic acid, o-Cresotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2086811
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96.0%

mp

163-165 °C (lit.)

solubility

methanol: soluble 5%, clear to slightly hazy (colorlessto faint brown)

SMILES string

Cc1cccc(C(O)=O)c1O

InChI

1S/C8H8O3/c1-5-3-2-4-6(7(5)9)8(10)11/h2-4,9H,1H3,(H,10,11)

InChI key

WHSXTWFYRGOBGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylsalicylic acid reacts with WOCl4 to yield tungsten(VI) oxo-salicylate complexes[1].

Application

3-Methylsalicylic acid was used in the synthesis of:
  • 8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one[2]
  • 7-hydroxy-8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one[2]
  • (R)-, (S)- and (RS)-hydroxymethylmexiletine[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Warren B Cross et al.
Dalton transactions (Cambridge, England : 2003), (7)(7), 1287-1293 (2005-03-23)
Tungsten(VI) oxo-salicylate complexes were prepared in moderate yield (47 to 63%) by the reactions of WOCl4 and two equivalents of either 3-methylsalicylic acid (MesaliH2) or 3,5-di-isopropylsalicylic acid (di-i-PrsaliH2). Performing the reaction in refluxing toluene afforded the two analogous ditungsten complexes
Krithika Delhiraja et al.
Environmental monitoring and assessment, 192(7), 432-432 (2020-06-17)
Emerging contaminants (ECs) have become an increasing area of concern due to the likely impacts of these compounds on human health and the environment. Generally, products which are used for households and personal care activities contribute to major percentage of
Synthesis of (R)-, (S)-, and (RS)-hydroxymethylmexiletine, one of the major metabolites of mexiletine.
Cavalluzzi MM, et al.
Tetrahedron Asymmetry, 18(20), 2409-2417 (2007)
Generalized Method for the Production of 1, 3-Benzoxazine, 1, 3-Benzothiazine, and Quinazoline Derivatives from 2-(Hydroxy, Thio, or Amino) Aromatic Acids Using Triphenylphosphine Thiocyanogen.
Pritchard KM, et al.
Synthetic Communications, 35(!2), 1601-1611 (2005)
Alena A Antipova et al.
Genome biology, 9(3), R47-R47 (2008-03-04)
Here we describe a proof-of-concept experiment designed to explore the possibility of using gene expression-based high-throughput screening (GE-HTS) to find inhibitors of a signaling cascade, using platelet derived growth factor receptor (PDGFR) signaling as the example. The previously unrecognized ability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service