Skip to Content
Merck
All Photos(1)

Key Documents

163651

Sigma-Aldrich

Thioacetanilide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CSNHC6H5
CAS Number:
Molecular Weight:
151.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

76-79 °C (lit.)

functional group

amine

SMILES string

CC(=S)Nc1ccccc1

InChI

1S/C8H9NS/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

MWCGLTCRJJFXKR-UHFFFAOYSA-N

General description

Metabolism and acute toxicity of thioacetanilide has been studied in rat. Thioacetanilide undergoes nucleophilic addition reaction with superoxide ion in dimethyl sulfoxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Michta et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 8), o411-o413 (2008-08-07)
The title compound, C(8)H(9)NS, has four symmetry-independent molecules in the asymmetric unit. These molecules link into two independent infinite N-H...S hydrogen-bonded chains in the a-axis direction with graph-set notation C(2)(2)(8). The NH-CS group adopts a trans conformation and forms a
Ester Muraglia et al.
Bioorganic & medicinal chemistry letters, 16(10), 2748-2752 (2006-03-01)
A series of aryltetrazolylacetanilides was synthesized and evaluated as HIV-1 non-nucleoside reverse transcriptase inhibitors on wild-type virus and on the clinically relevant K103N mutant strain. Extensive SAR investigation led to potent compounds, with nanomolar activity on K103N, and orally bioavailable
Peng Zhan et al.
Bioorganic & medicinal chemistry, 17(16), 5775-5781 (2009-08-01)
A series of 2-(1-aryl-1H-imidazol-2-ylthio)acetamide [imidazole thioacetanilide (ITA)] derivatives were synthesized and evaluated as potent inhibitors of human immunodeficiency virus type-1 (HIV-1). Among them, the most potent HIV-1 inhibitors were 4a5 (EC(50)=0.18microM), and 4a2 (EC(50)=0.20microM), which were more effective than the
Li-Rong Wen et al.
The Journal of organic chemistry, 73(5), 1852-1863 (2008-01-29)
A series of unusual fused tricyclic thiochromeno[2,3-b]pyridines were successfully synthesized by tandem [3 + 3] annulation and SNAr of beta-(2-chloroaroyl) thioacetanilides with activated 4-arylidene-2-phenyloxazol-5(4H)-ones or aromatic aldehydes and ethyl 2-cyanoacetate under microwave irradiation, respectively. Because of the existence of the
Reactivity of superoxide ion with thioamides in dimethyl sulfoxide.
Paez OA, et al.
The Journal of Organic Chemistry, 53(10), 2166-2170 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service