Skip to Content
Merck
All Photos(1)

Key Documents

161977

Sigma-Aldrich

N-Cyclohexylisopropylamine

98%

Synonym(s):

N-Isopropylcyclohexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHC6H11
CAS Number:
Molecular Weight:
141.25
Beilstein:
2070639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

60-65 °C/12 mmHg (lit.)

density

0.859 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)NC1CCCCC1

InChI

1S/C9H19N/c1-8(2)10-9-6-4-3-5-7-9/h8-10H,3-7H2,1-2H3

InChI key

UYYCVBASZNFFRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Cyclohexylisopropylamine was used as reagent in the synthesis of 7-hydroxy cannabidiol and cannabidiol-7-oic acid.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ahmed A Noser et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-22)
A series of novel quinazolinone derivatives (2-13) was synthesized and examined for their cytotoxicity to HepG2, MCF-7, and Caco-2 in an MTT assay. Among these derivatives, compounds 4 and 9 exhibited significant cytotoxic activity against Caco-2, HepG2, and MCF-7 cancer
Raphael Mechoulam et al.
Chemistry and physics of lipids, 121(1-2), 35-43 (2002-12-31)
Over the last few years considerable attention has focused on cannabidiol (CBD), a major non-psychotropic constituent of Cannabis. In Part I of this review we present a condensed survey of the chemistry of CBD; in Part II, to be published

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service