Skip to Content
Merck
All Photos(2)

Key Documents

161705

Sigma-Aldrich

Di-2-pyridyl ketone oxime

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N3O
CAS Number:
Molecular Weight:
199.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

142-143 °C (lit.)

functional group

amine
oxime

storage temp.

2-8°C

SMILES string

O\N=C(\c1ccccn1)c2ccccn2

InChI

1S/C11H9N3O/c15-14-11(9-5-1-3-7-12-9)10-6-2-4-8-13-10/h1-8,15H

InChI key

KLOQKBNABUIDIU-UHFFFAOYSA-N

General description

Di-2-pyridyl ketone oxime forms hexanuclear clusters with extremely rare manganese oxidation-state combinations[1]. It forms neutral binuclear and cationic trinuclear, pentanuclear or hexanuclear complexes in copper chemistry[2].

Application

Di-2-pyridyl ketone oxime was used as a standard in the metal assay for determination of cobalt[3]. It may be employed as metal-complexing agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constantinos J Milios et al.
Dalton transactions (Cambridge, England : 2003), (3)(3), 501-511 (2005-01-27)
The use of di-2-pyridyl ketone oxime, (py)2CNOH, in manganese carboxylate chemistry has been investigated. Using a variety of synthetic routes complexes [Mn(O2CPh)2{(py)2CNOH}2].0.25H2O (1.0.25H2O), Mn4(O2CPh)2{(py)2CO2}2{(py)2CNO}2Br2].MeCN (2.MeCN), [Mn4(O2CPh)2{(py)2CO2}2{(py)2CNO}2Cl(2)].2MeCN (3.2MeCN), [Mn4(O2CMe)2{(py)2CO2}2{(py)2CNO}2Br2].2MeCN (4.2MeCN), [Mn4(O2CMe)2{(py)2CO2}2{(py)2CNO}2(NO3)2].MeCN.H2O (5.MeCN.H2O) and [Mn2(O2CCF3)2(hfac)2{(py)2CNOH}2] (6) have been isolated in good yields.
Tereza Afrati et al.
Dalton transactions (Cambridge, England : 2003), (25)(25), 2658-2668 (2007-06-20)
The use of di-2-pyridyl ketone oxime (Hpko)/X- "blends" (X- = OH-, Cl-, ClO4-) in copper chemistry has yielded neutral binuclear and cationic trinuclear, pentanuclear or hexanuclear complexes. Various synthetic procedures have led to the synthesis of compounds [Cu5(pko)7].[ClO4]3.2CH3OH.2H2O (1), [Cu3(pko)3(OH)(Cl)]2[Ph4B]2.4DMF.2H2O
Paul V Bernhardt et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 8(8), 866-880 (2003-10-18)
Di-2-pyridyl ketone isonicotinoyl hydrazone (HPKIH) and a range of its analogues comprise a series of monobasic acids that are capable of binding iron (Fe) as tridentate ( N, N, O) ligands. Recently, we have shown that these chelators are highly
Constantinos C Stoumpos et al.
Inorganic chemistry, 49(10), 4388-4390 (2010-04-16)
The use of phenyl-2-pyridyl ketone oxime and di-2-pyridyl ketone oxime in Mn chemistry has led to hexanuclear clusters with unprecedented (Mn(II)(4)Mn(III)Mn(IV)) or extremely rare (Mn(II)Mn(III)(5) and Mn(II)(3)Mn(III)(3)) metal oxidation-state combinations and uncommon structural motifs.
M W Reeves et al.
Journal of clinical microbiology, 14(1), 48-54 (1981-07-01)
A total of 13 egg lecithins, 12 beef heart lecithins, and 15 beef heart cardiolipins were assayed for the ability to function in the Venereal Disease Research Laboratory microflocculation test, as well as for purity, fatty acid composition, free amines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service