Skip to Content
Merck
All Photos(1)

Key Documents

16006

Sigma-Aldrich

SP Brij® S2 MBAL

main component: diethylene glycol octadecyl ether

Synonym(s):

Brij® S2, Polyethylene glycol octadecyl ether, Polyoxyethylene (2) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H37(OCH2CH2)2OH
CAS Number:
Molecular Weight:
358.60
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

description

non-ionic

mol wt

358.60 g/mol

technique(s)

protein quantification: suitable

mp

44-45 °C (lit.)

HLB value

4.9

density

0.893 g/mL at 25 °C (lit.)

application(s)

microbiology

SMILES string

O(CC[O])CCCCCCCCCCCCCCCCCC

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J P Lens et al.
Journal of biomaterials science. Polymer edition, 8(12), 963-982 (1997-01-01)
Alkyl-poly(ethylene oxide) (PEO) surfactants containing a terminal hydroxyl, sulfate, or carboxylate group were grafted at the surface of poly(ethylene) (PE) samples to improve their blood compatibility. Grafting was achieved by immobilizing PEO surfactants on PE using an argon plasma treatment.
Anne Huhtala et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 19(1), 11-21 (2003-03-22)
This study was undertaken to investigate the use of the in vitro test WST-1, an assay of cell proliferation and viability, for a preliminary safety evaluation of topical ophthalmic preparations. The cytotoxicity of two surfactants, benzalkonium chloride (BAC) and polyoxyethylene-20-stearyl
Y C Lee et al.
Journal of pharmaceutical sciences, 86(4), 430-433 (1997-04-01)
An ocular insert is developed for the controlled systemic delivery of insulin. Commercially available Gelfoam absorbable gelatin sponge, USP, is used in the fabrication of the ocular insert in the form of a matrix system. Two eyedrop formulations and 13
N Albertson et al.
Journal of clinical microbiology, 36(5), 1232-1235 (1998-05-09)
The gastrointestinal pathogen Helicobacter pylori requires supplementation with either fetal calf serum (FCS), bovine serum albumin (BSA), or (2,6-dimethyl)-beta-cyclodextrin (CD) for growth in a complex or defined medium. Because the availability of medium in which all components were chemically defined
J Q Zhang et al.
Drug delivery, 14(6), 381-387 (2007-08-19)
The study describes the development of stealth solid lipid nanoparticles (SLNs) as colloidal carriers for silibinin, a drug with very low solubility. Stealth SLNs were constituted mainly of bioacceptable and biodegradable lipids, such as stearic acid and surfactant Brij 78

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service