Skip to Content
Merck
All Photos(1)

Key Documents

159883

Sigma-Aldrich

2-Methoxybenzylamine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€32.50
25 G
€86.40

€32.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€32.50
25 G
€86.40

About This Item

Linear Formula:
CH3OC6H4CH2NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
1100448
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€32.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

227 °C/724 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

COc1ccccc1CN

InChI

1S/C8H11NO/c1-10-8-5-3-2-4-7(8)6-9/h2-5H,6,9H2,1H3

InChI key

PXJACNDVRNAFHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Methoxybenzylamine was used in the preparation of 6-substituted purines.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Zatloukal et al.
European journal of medicinal chemistry, 61, 61-72 (2012-07-10)
Several inhibitors of cyclin-dependent kinases (CDKs), including the 2,6,9-trisubstituted purine derivative roscovitine, are currently being evaluated in clinical trials as potential anticancer drugs. Here, we describe a new series of roscovitine derivatives that show increased potency in vitro. The series

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service