Skip to Content
Merck
All Photos(2)

Key Documents

155888

Sigma-Aldrich

trans-2-Methyl-3-phenyl-2-propen-1-ol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(CH3)CH2OH
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.572 (lit.)

bp

77 °C/0.1 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

[H]\C(=C(\C)CO)c1ccccc1

InChI

1S/C10H12O/c1-9(8-11)7-10-5-3-2-4-6-10/h2-7,11H,8H2,1H3/b9-7+

InChI key

LLNAMUJRIZIXHF-VQHVLOKHSA-N

Application

trans-2-Methyl-3-phenyl-2-propen-1-ol was used in the preparation of 5-methyl-4-phenyl-5-hexen-2-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Angermaier et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 362(1), 33-38 (1981-01-01)
With ethanol as electron donor, resting cells of Clostridium kluyveri reduced (E)-2-methyl-2-buten-1-ol, (E)-3-methyl-2-penten-1-ol and (E)-2-methyl-3-phenyl-2-propen-1-ol to (R)-2-methyl-1-butanol, (R)-3-methyl-1-1-pentanol and (R)-2-methyl-3-phenyl-1-propanol, respectively. Within the experimental errors of ORD measurements the products were optically pure. That means that only one of the
Masao Morita et al.
The Journal of organic chemistry, 71(16), 6285-6286 (2006-07-29)
One-pot synthesis of gamma,delta-unsaturated carbonyl compounds from allyl alcohols and vinyl or isopropenyl acetates was achieved through in situ generation of allyl vinyl ethers by the action of the [IrCl(cod)]2 complex followed by Claisen rearrangement of the resulting ethers. For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service