Skip to Content
Merck
All Photos(1)

Key Documents

153176

Sigma-Aldrich

2-Phenoxybenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4CO2H
CAS Number:
Molecular Weight:
214.22
Beilstein:
978452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

110-112 °C (lit.)

functional group

carboxylic acid
phenoxy

SMILES string

OC(=O)c1ccccc1Oc2ccccc2

InChI

1S/C13H10O3/c14-13(15)11-8-4-5-9-12(11)16-10-6-2-1-3-7-10/h1-9H,(H,14,15)

InChI key

PKRSYEPBQPFNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Phenoxybenzoic acid was used as an internal standard in the quantification of pyrethroid metabolites[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nor Saadah Mohd Yusof
Ultrasonics sonochemistry, 71, 105360-105360 (2020-10-31)
The ion exchange constant, KXBr (for the case of cetyltrimethylammonium bromide, CTABr, in this study) is a method dependant characterization of ion exchange process by counterions, X and Br with different relative binding ratios. In this report, the ion exchange
Yuko Ueda et al.
Analytical and bioanalytical chemistry, 410(24), 6207-6217 (2018-07-27)
Four pyrethroids (PYRs), metofluthrin, profluthrin, tefluthrin, and transfluthrin, which were newly developed and have relatively high vapor activity at ambient temperature, are now playing a key role in safely controlling insects in our daily lives. We developed a sensitive and
T Schettgen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 778(1-2), 121-130 (2002-10-12)
We have developed and validated a new, reliable and very sensitive method for the determination of the urinary metabolites of the most common pyrethroids in one analytical run. After acidic hydrolysis for the cleavage of conjugates, the analytes cis-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic acid
Sayyed Abbas Tabatabai et al.
Iranian journal of pharmaceutical research : IJPR, 12(Suppl), 105-111 (2013-11-20)
Benzodiazepines are useful drugs for treatment of sleep disorders, anxiety, seizure cases and skeletal muscle cramps. Some derivatives of 2-(2-Phenoxy) phenyl-1, 3, 4-oxadiazole were synthesized as benzodiazepine receptor agonists. Conformational analysis and superimposition of energy minima conformers of the compounds
Yuan Liu et al.
Applied microbiology and biotechnology, 104(17), 7345-7354 (2020-07-16)
Pyrethroids are insecticides that are widely used in rural and urban areas worldwide. After entering the environment, pyrethroids are rapidly metabolized or degraded by various biological or abiotic methods. In this study, a single-chain variable fragment (scFv) which could simultaneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service