Skip to Content
Merck
All Photos(1)

Documents

151564

Sigma-Aldrich

5-Hydroxydopamine hydrochloride

98%

Synonym(s):

3,4,5-Trihydroxyphenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
205.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

218-220 °C (lit.)

SMILES string

Cl.NCCc1cc(O)c(O)c(O)c1

InChI

1S/C8H11NO3.ClH/c9-2-1-5-3-6(10)8(12)7(11)4-5;/h3-4,10-12H,1-2,9H2;1H

InChI key

WSZSSLCEWCKAKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Hydroxydopamine hydrochloride was used in the preparation of hydrogen peroxide, superoxide radical and hydroxyl radical.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jose A Rodríguez-Gómez et al.
Stem cells (Dayton, Ohio), 25(4), 918-928 (2006-12-16)
The derivation of dopamine neurons is one of the best examples of the clinical potential of embryonic stem (ES) cells, but the long-term function of the grafted neurons has not been established. Here, we show that, after transplantation into an
F Neff et al.
Annals of the New York Academy of Sciences, 973, 70-74 (2002-12-18)
We show that the glial cell line-derived neurotrophic factor (GDNF) activates the PI3K/Akt-signaling pathway in human neuroblastoma cells that express functional Ret-receptor complexes. Consistent with this finding we show PI3K-dependent Bad-inactivation by binding to 14-3-3 proteins in response to GDNF.
E Kienzl et al.
Life sciences, 65(18-19), 1973-1976 (1999-11-27)
The mechanisms leading to degeneration of melanized dopaminergic neurons in the brain stem, and particularly in the substantia nigra zona compacta (SNZC) in patients with Parkinson's disease (PD) are still unknown. Demonstration of increased iron Fe(III) in SNZC of PD
C Spenger et al.
Experimental brain research, 112(1), 47-57 (1996-11-01)
Free-floating roller tube cultures of human fetal (embryonic age 6-10 weeks post-conception) and rat fetal (embryonic day 13) ventral mesencephalon were prepared. After 7-15 days in vitro, the mesencephalic tissue cultures were transplanted into the striatum of adult rats that
K Jellinger et al.
Journal of neural transmission. Supplementum, 46, 297-314 (1995-01-01)
The presence of 5-Hydroxydopamine (5-OHDA) and 6-Hydroxydopamine (6-OHDA) in the urine of parkinsonian patients on levodopa medication was reported by Andrew et al. (1993). To answer the question about the putative relevance of 6-OHDA endogenously formed in the brain for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service