Skip to Content
Merck
All Photos(1)

Key Documents

14972

Sigma-Aldrich

Boc-8-Aoc-OH

≥97.0%

Synonym(s):

8-(Boc-amino)caprylic acid, 8-(Boc-amino)octanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNH(CH2)7CO2H
CAS Number:
Molecular Weight:
259.34
Beilstein:
2268966
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0%

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

56-59 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NCCCCCCCC(O)=O

InChI

1S/C13H25NO4/c1-13(2,3)18-12(17)14-10-8-6-4-5-7-9-11(15)16/h4-10H2,1-3H3,(H,14,17)(H,15,16)

InChI key

FPRZYWCRQHFPSX-UHFFFAOYSA-N

Other Notes

Reagent to introduce a protected ω-amino alkyl spacer. Longer spacer arms often impart increased activity to the substrate.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

B J Bradbury et al.
Journal of medicinal chemistry, 33(2), 741-748 (1990-02-01)
A functionalized congener approach was used to design ligands for muscarinic cholinergic receptors (mAChRs). A series of omega-functionalized alkyl amides of N-methyl-4-(1-pyrrolidinyl)-2-butynamine (22) were prepared as functionalized analogues of UH 5 [N-methyl-N-[4-(1-pyrrolidinyl)-2-butynyl]acetamide], a muscarinic agonist related to oxotremorine. Intermediate 22

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service