Skip to Content
Merck
All Photos(1)

Key Documents

145807

Sigma-Aldrich

2-Bromopropionyl chloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHBrCOCl
CAS Number:
Molecular Weight:
171.42
Beilstein:
773800
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid

grade

technical grade

form

liquid

refractive index

n20/D 1.477 (lit.)

bp

131-133 °C (lit.)

density

1.7 g/mL at 25 °C (lit.)

SMILES string

CC(Br)C(Cl)=O

InChI

1S/C3H4BrClO/c1-2(4)3(5)6/h2H,1H3

InChI key

OZGMODDEIHYPRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromopropionyl chloride was used to synthesize cellulose macroinitiator for homogeneous atom transfer radical polymerization[1]. It was used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase synthesis of C-terminal peptide amides using a photoremovable a-methylphenacylamido anchoring linkage.
Ajayaghosh A and Pillai VNR.
Proceedings of the Indian Academy of Sciences - Section A, 100(5), 389-396 (1988)
Xiaofeng Sui et al.
Biomacromolecules, 9(10), 2615-2620 (2008-09-09)
Cellulose-graft-poly(N,N-dimethylamino-2-ethyl methacrylate) (cellulose-g-PDMAEMA) copolymers were prepared by homogeneous atom transfer radical polymerization (ATRP) under mild conditions. Cellulose macroinitiator was successfully synthesized by direct acylation of cellulose with 2-bromopropionyl bromide in a room temperature ionic liquid (RTIL), 1-allyl-3-methylimidazolium chloride. Copolymers were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service