Skip to Content
Merck
All Photos(1)

Key Documents

145653

Sigma-Aldrich

3-Methoxy-2(1H)-pyridone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

115-117 °C (lit.)

SMILES string

COC1=CC=CNC1=O

InChI

1S/C6H7NO2/c1-9-5-3-2-4-7-6(5)8/h2-4H,1H3,(H,7,8)

InChI key

LKIMDXQLHFCXQF-UHFFFAOYSA-N

Application

3-Methoxy-2(1H)-pyridone was used in synthesis of azabenzoisocoumarins by double ipso substitutions[1]. It was used as the starting reagent during the total synthesis of cepabactin[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cédric Klumpp et al.
Bioorganic & medicinal chemistry letters, 15(6), 1721-1724 (2005-03-05)
A novel and straightforward total synthesis of cepabactin and its iron (III) complex is described. The latter compound was compared and identified to that obtained from the cultures of Burkholderia cepacia. On treatment of the growth medium of two different
Free radical reactions for heterocycle synthesis. Part 6: 2-Bromobenzoic acids as building blocks in the construction of nitrogen heterocycles.
Zhang W and Pugh G.
Tetrahedron, 59(17), 3009-3018 (2003)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service