Skip to Content
Merck
All Photos(1)

Documents

145467

Sigma-Aldrich

2-Chlorobenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2OH
CAS Number:
Molecular Weight:
142.58
Beilstein:
2042182
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

227 °C (lit.)

mp

69-71 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, colorless

SMILES string

OCc1ccccc1Cl

InChI

1S/C7H7ClO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2

InChI key

MBYQPPXEXWRMQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Major urinary metabolites of 2-chlorobenzyl alcohol in rats were 2-chlorohippuric acid, 2-chlorobenzyl cysteine, 2-chlorobenzoic acid and 2-chlorobenzyl glucuronic acid. It is a possible intermediate formed during the metabolism of 2-chloromandelic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Brewster et al.
Xenobiotica; the fate of foreign compounds in biological systems, 17(8), 911-924 (1987-08-01)
1. The fate of 3H-ring labelled, 14C-cyanide labelled and (14C = C) side chain labelled 2-chlorobenzylidene malononitrile (CS), 2-chlorobenzyl alcohol and 2-chlorobenzyl malononitrile (dihydro CS) in rats and isolated rat liver cells has been examined. 2. CS was administered both
G S J Mannens et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(4), 554-565 (2006-08-29)
RWJ-333369 (1,2-ethanediol, [1-2-chlorophenyl]-, 2-carbamate, [S]-; CAS Registry Number 194085-75-1) is a novel neuromodulator in clinical development for the treatment of epilepsy. To study the disposition of RWJ-333369, eight healthy male subjects received a single oral dose of 500 mg of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service