Skip to Content
Merck
All Photos(1)

Key Documents

144916

Sigma-Aldrich

2-Amino-5-methylphenol

98%

Synonym(s):

2-Hydroxy-4-methylaniline, 4-Amino-3-hydroxytoluene, 6-Amino-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Beilstein:
386144
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
98%

Assay

98%

form

powder

mp

159-162 °C (lit.)

SMILES string

Cc1ccc(N)c(O)c1

InChI

1S/C7H9NO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,8H2,1H3

InChI key

HCPJEHJGFKWRFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-5-methylphenol reacts with bovine hemoglobin to form 2-amino-4,4α-dihydro-4α-7-dimethyl-3H-phenoxazine-3-one, which inhibits the proliferation of Poliovirus in Vero cells. It is converted to dihydrophenoxazinone by purified human hemoglobin.

Application

2-Amino-5-methylphenol was used in the synthesis of tridentate Schiff base ligand and novel non-metallocene catalysts with phenoxy-imine ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Akiko Iwata et al.
The Tohoku journal of experimental medicine, 200(3), 161-165 (2003-10-03)
2-Amino-4,4alpha-dihydro-4alpha-7-dimethyl-3H-phenoxazine-3-one (Phx), which was produced by the reaction of bovine hemoglobin with 2-amino-5-methylphenol, inhibited the proliferation of poliovirus in Vero cells between 0.25 microg/ml and 2 microg/ml with maximal antiviral acitivity at 1 microg/ml. These results suggest that Phx may
The investigation of novel non-metallocene catalysts with phenoxy-imine ligands for ethylene (co-) polymerization.
Zhang X, et al.
Polymer International, 62(3), 419-426 (2012)
Synthesis,structural characterization and catalytic activity study of Mn(II), Fe(III), Ni(II), Cu(II) and Zn(II) complexes of quinoxaline-2-carboxalidine-2-amino-5-methylphenol: Crystal structure of the nickel (II) complex.
Sebastian M, et al.
Polyhedron, 29(15), 3014-3020 (2010)
A Tomoda et al.
Biochimica et biophysica acta, 1117(3), 306-314 (1992-10-27)
We found that 2-amino-5-methylphenol was converted to the dihydrophenoxazinone with a reddish brown color by purified human hemoglobin, lysates of human erythrocytes, and human erythrocytes. The reddish brown compound was identified as 2-amino-4,4 alpha-dihydro-4 alpha,7-dimethyl-3H-phenoxazin-3-one by the measurement of NMR
A Tomoda et al.
Bioorganic & medicinal chemistry letters, 11(8), 1057-1058 (2001-05-01)
A simple and rapid preparation method for a novel antitumor agent, 2-amino-4,4a-dihydro-4a,7-dimethyl-3H-phenoxazine-3-one (Phx) was described. The procedure included (1) the reaction of bovine hemolysates with 2-amino-5-methylphenol, (2) one-shot denaturation of hemoglobin and proteins by methanol, and removal of the denatured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service