Skip to Content
Merck
All Photos(1)

Documents

144886

Sigma-Aldrich

3-(Methylthio)aniline

97%

Synonym(s):

3-(Methylmercapto)aniline, 3-Aminothioanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SC6H4NH2
CAS Number:
Molecular Weight:
139.22
Beilstein:
2078599
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.637 (lit.)

bp

163-165 °C/16 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

SMILES string

CSc1cccc(N)c1

InChI

1S/C7H9NS/c1-9-7-4-2-3-6(8)5-7/h2-5H,8H2,1H3

InChI key

KCHLDNLIJVSRPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-(Methylthio)aniline was used in the synthesis of phenyl azobenzene sulfonamide derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Turan Gul et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(8), 1270-1276 (2016-03-18)
Mammalian flavin-containing monooxygenases, which are difficult to obtain and study, play a major role in detoxifying various xenobiotics. To provide alternative biocatalytic tools to generate flavin-containing monooxygenases (FMO)-derived drug metabolites, a collection of microbial flavoprotein monooxygenases, sequence-related to human FMOs
Takashi Ikawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(19), 4320-4332 (2020-01-03)
Benzynes were selectively generated in situ from phenols and trapped regioselectively with potassium hexamethyldisilazide to form primary anilines following acidic workup. The direct conversion of a phenolic hydroxyl group into a free amino group is a useful method for the
Wei-Jern Tsai et al.
Bioorganic & medicinal chemistry letters, 16(17), 4440-4443 (2006-07-04)
A series of phenylazobenzenesulfonamide derivatives were designed and synthesized for the evaluation as selective cyclooxygenase-2 (COX-2) inhibitors in a cellular assay using human whole blood (HWB) and an enzymatic assay using purified ovine enzymes. Extensive structure-activity relationships (SAR) were studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service