Skip to Content
Merck
All Photos(1)

Key Documents

13611

Sigma-Aldrich

Wang resin

extent of labeling: ~1.0 mmol/g loading

Synonym(s):

p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~1.0 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

application(s)

peptide synthesis

functional group

alcohol

Looking for similar products? Visit Product Comparison Guide

Other Notes

Support used in solid phase peptide synthesis; Suppression of side reactions during the attachement of Fmoc-amino acids to the Wang resin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.W. van Nispen et al.
Rec. Trav. Chim., 104, 99-99 (1985)
S.S. Wang
Journal of the American Chemical Society, 95, 13128-13128 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service