Skip to Content
Merck
All Photos(1)

Key Documents

133124

Sigma-Aldrich

Trichloroethylene

reagent grade, ≥99.0%, contains ~1% 1,2-epoxybutane as inhibitor

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

reagent grade

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

770 °F

contains

~1% 1,2-epoxybutane as inhibitor

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive dechlorination of trichloroethylene (TCE) in competition with Fe and Mn oxides-observed dynamics in H2-dependent terminal electron accepting processes.
Paul L, et al.
Geomicrobiology Journal (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 332-332 (1994)
T M Vogel et al.
Applied and environmental microbiology, 49(5), 1080-1083 (1985-05-01)
Tetrachloroethylene (PCE) and trichloroethylene (TCE), common industrial solvents, are among the most frequent contaminants found in groundwater supplies. Due to the potential toxicity and carcinogenicity of chlorinated ethylenes, knowledge about their transformation potential is important in evaluating their environmental fate.
Eagleson M.
Concise Encyclopedia Chemistry, 215-215 (1994)
Jo Philips et al.
Chemosphere, 91(1), 7-13 (2012-12-12)
Electron donor limitations likely reduce microbial enhanced trichloroethene (TCE) dense non-aqueous phase liquid (DNAPL) dissolution. This study quantitatively examined the relation between the DNAPL dissolution enhancement and the electron donor supply rate. An experiment used diffusion-cells with a 5.5 cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service