132543
4-Bromobutyl phenyl ether
97%
Synonym(s):
(4-Bromobutoxy)benzene, 4-Phenoxybutyl bromide, Phenoxybutyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
bp
153-156 °C/18 mmHg (lit.)
mp
41-43 °C (lit.)
SMILES string
BrCCCCOc1ccccc1
InChI
1S/C10H13BrO/c11-8-4-5-9-12-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2
InChI key
QBLISOIWPZSVIK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemistry, an Asian journal, 3(8-9), 1472-1478 (2008-07-10)
Regioselective double alkylation of styrenes with alkyl Grignard reagents and alkyl bromides having a heteroatom functional group at the beta-position has been achieved by the use of a titanocene catalyst in THF. When ether was used instead of THF as
Electrochemical reduction of 1, 4-dihalobutanes at carbon cathodes in dimethylformamide.
Journal of Electroanalytical Chemistry, 380(1), 147-160 (1995)
The Journal of organic chemistry, 76(7), 2355-2359 (2011-03-11)
An experimentally simple sequential one-pot RuAAC reaction, affording 1,5-disubstituted 1H-1,2,3-triazoles in good to excellent yields starting from an alkyl halide, sodium azide, and an alkyne, is reported. The organic azide is formed in situ by treating the primary alkyl halide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service