Skip to Content
Merck
All Photos(1)

Key Documents

132152

Sigma-Aldrich

2,4,5-Trimethoxybenzaldehyde

98%

Synonym(s):

Asaronaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CHO
CAS Number:
Molecular Weight:
196.20
Beilstein:
1951403
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 132152, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

mp

112-114 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cc(OC)c(OC)cc1OC

InChI

1S/C10H12O4/c1-12-8-5-10(14-3)9(13-2)4-7(8)6-11/h4-6H,1-3H3

InChI key

IAJBQAYHSQIQRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D Dillon et al.
Mutagenesis, 13(1), 19-26 (1998-03-10)
Several aldehydes and peroxides were tested for mutagenicity using Salmonella typhimurium tester strains TA97a, TA100, TA102 and TA104, in the presence and absence of Aroclor-induced liver S9 mix from F344 rats and B6C3F1 mice, in either preincubation or vapour phase
Chien-Chi Chiang et al.
Journal of agricultural and food chemistry, 61(38), 9160-9165 (2013-08-13)
The antiproliferation activity of the ethanol extract of A. cinnamomea mycelium on hepatocellular cancer cells HepG2 was found to be associated with aroma intensity of the broth during fermentation. We hypothesized that some of the volatile compounds are the precursors
Xiao-Gang Tong et al.
Journal of natural products, 73(6), 1160-1163 (2010-05-19)
A new cadinane-type sesquiterpenoid, tatarinowin A (1), two phenylpropanoids, tatarinoids A (2) and B (3), and a trinorlignan, tatarinoid C (4), along with 15 known compounds including two pairs of mixtures were isolated from the rhizome of Acorus tatarinowii. The
C-C Chen et al.
Letters in applied microbiology, 44(4), 387-392 (2007-04-03)
To investigate the active ingredient in fruiting bodies and to produce it with cultured mycelium in Antrodia camphorata (BCRC 35398). The volatile components from the fruiting bodies, the liquid cultured broth of A. camphorata and Cinnamomum kanehirae wood were separately
X Xu et al.
Infection and immunity, 69(7), 4465-4472 (2001-06-13)
Treponema denticola does not appear to produce siderophores, so it must acquire iron by other pathways. Indeed, T. denticola has been shown to have an iron-regulated 44-kDa outer membrane protein (HbpA) with hemin binding ability. To characterize the HbpA protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service