Skip to Content
Merck
All Photos(1)

Key Documents

130826

Sigma-Aldrich

3-Acetoxypyridine

99%

Synonym(s):

3-Pyridyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

refractive index

n20/D 1.503 (lit.)

bp

92 °C/9 mmHg (lit.)

density

1.141 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1cccnc1

InChI

1S/C7H7NO2/c1-6(9)10-7-3-2-4-8-5-7/h2-5H,1H3

InChI key

QZDWODWEESGPLC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The addition of acetic acid to pyridynes and quinolynes.
Fleet GWJ et al.
J. Chem. Soc. Sect. C, 3948-3950 (1971)
Dao Feng Xiang et al.
Biochemistry, 53(21), 3476-3485 (2014-05-17)
The uncharacterized protein Rsp3690 from Rhodobacter sphaeroides is a member of the amidohydrolase superfamily of enzymes. In this investigation the gene for Rsp3690 was expressed in Escherichia coli and purified to homogeneity, and the three-dimensional structure was determined to a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service