Skip to Content
Merck
All Photos(1)

Key Documents

13032

Sigma-Aldrich

2-(Benzoyloxymethyl)benzoyl chloride

≥97.0% (AT)

Synonym(s):

2-(Hydroxymethyl)benzoyl chloride benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO3
CAS Number:
Molecular Weight:
274.70
Beilstein:
2131346
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (AT)

form

solid

contains

~1% CaCO3 as stabilizer

mp

59-61 °C

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccccc1COC(=O)c2ccccc2

InChI

1S/C15H11ClO3/c16-14(17)13-9-5-4-8-12(13)10-19-15(18)11-6-2-1-3-7-11/h1-9H,10H2

InChI key

UDEQFYWNFHWPRH-UHFFFAOYSA-N

Application

2-(Benzoyloxymethyl)benzoyl chloride may be used in chemical synthesis studies.

Other Notes

Reagent for a novel amine protection which gives amides the lability of esters

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B.F. Cain
The Journal of Organic Chemistry, 41, 2029-2029 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service