Skip to Content
Merck
All Photos(1)

Key Documents

127906

Sigma-Aldrich

5-Amino-1,3,4-thiadiazole-2-thiol

95%

Synonym(s):

2-Amino-5-mercapto-1,3,4-thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3N3S2
CAS Number:
Molecular Weight:
133.20
Beilstein:
81728
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

235 °C (dec.) (lit.)

SMILES string

Nc1nnc(S)s1

InChI

1S/C2H3N3S2/c3-1-4-5-2(6)7-1/h(H2,3,4)(H,5,6)

InChI key

GDGIVSREGUOIJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-amino-1,3,4-thiadiazole-2-thiol was used to prepare new amines exhibiting anti-convulsant activity[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Wei Lee et al.
Biosensors & bioelectronics, 133, 215-222 (2019-04-06)
Early diagnosis of bacterial infections is crucial to improving survival rates by enabling treatment with appropriate antibiotics within the first few hours of infection. This paper presents a highly sensitive amperometric biosensor for the detection of several pathogenic bacterial cells
M Kavlakova et al.
Arzneimittel-Forschung, 62(12), 599-602 (2012-10-25)
The coordination properties of Pt(II) with 5-amino-1,3,4-thiadiazole-2-thiol [CAS 2349-67-9] (L 1 ) and its novel violurate adduct (L 2 ), both in solution and in solid state, are studied by means of conventional IR-spectroscopy, single crystal X-ray diffraction and thermal
Rodica Olar et al.
Materials (Basel, Switzerland), 13(20) (2020-10-22)
The derivatives 5,7-dimethyl[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate (1) and 7-methyl-5-phenyl[1,3,4]thiadiazolo[3,2-a]pyrimidin-4-ium-2-thiolate (2) were fully characterized by single-crystal X-ray diffraction. Their supramolecular structure is built through both π-π stacking and C=S-π interactions for both compounds. The embedment of the tested compounds into Fe3O4@C18 core-shell nanocoatings increased
Mohammad Yusuf et al.
Chemical biology & drug design, 81(5), 666-673 (2013-01-31)
New imines, derived from aromatic aldehyde, chalcones and 5-amino-1,3,4-thiadiazole-2-thiol exhibited promising anti-convulsant activity which is explained through chemo-biological interactions at receptor site producing the inhibition of human Carbonic Anhydrase-II enzyme (hCA-II) through the proposed pharmacophore model at molecular levels as
Mohammad Hossein Ghanbari et al.
Mikrochimica acta, 186(9), 641-641 (2019-08-25)
A nanocomposite was prepared with reduced graphene oxide, gold nanoparticles and an electropolymerized film made from 2-amino-5-mercapto-1,3,4-thiadiazole. An electrochemical sensor for doxorubicin (DOX) was constructed by modifying a glassy carbon electrode (GCE) with the nanocomposite. The modified GCE was studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service