Skip to Content
Merck
All Photos(3)

Key Documents

124826

Sigma-Aldrich

4-Cyanobenzoyl chloride

4-Cyanobenzoyl chloride

98%

Synonym(s):

4-Cyanobenzoic acid chloride, p-Cyanobenzoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€26.40
5 G
€52.40
25 G
€143.00

€26.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€26.40
5 G
€52.40
25 G
€143.00

About This Item

Linear Formula:
NCC6H4COCl
CAS Number:
Molecular Weight:
165.58
Beilstein:
386729
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€26.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

68-70 °C (lit.)

functional group

acyl chloride
nitrile

SMILES string

ClC(=O)c1ccc(cc1)C#N

InChI

1S/C8H4ClNO/c9-8(11)7-3-1-6(5-10)2-4-7/h1-4H

InChI key

USEDMAWWQDFMFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Cyanobenzoyl chloride participates in the benzylamine acylation of Argopore MB-CHO resin in the presence of pyridine and catalyst[1]. It reacts with 4-hydroxy-2,2,6,6-tetramethyl-1-piperidinoxyl (4-hydroxy-TEMPO) in pyridine under nitrogen atmosphere to form 4-cyanobenzoyl-TEMPO[2].

Application

4-Cyanobenzoyl chloride has been used in the synthesis of new liquid crystalline heteroaromatic compounds containing the five-membered isoxazole, tetrazole and 1,2,4-oxadiazole rings[3]. It has been used in the synthesis of substituted benzoate esters and to study their excited-state behavior[4].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic field effects on the photoinduced electron transfer of 10-methylphenothiazine with 4-(4-cyanobenzoyloxy) TEMPO in fluid solutions.
Mori Y, et al.
Chemical Physics Letters, 286(5-6), 446-451 (1998)
Intramolecular electron transfer in the photochemistry of substituted 1-naphthylmethyl esters of benzoic acids.
DeCosta, DP and Pincock JA.
Canadian Journal of Chemistry, 70(7), 1879-1885 (1992)
Synthesis of liquid crystals materials derived from oxadiazole, isoxazole and tetrazole heterocycles.
dos Santos DR, et al.
ARKIVOC (Gainesville, FL, United States), 17, 157-166 (2008)
[2 patients with intestinal perforation following coagulation of the tubes by means of laparoscopy].
F J van Assen et al.
Nederlands tijdschrift voor geneeskunde, 119(8), 304-306 (1975-02-22)
Md Najbul Hoque et al.
Dalton transactions (Cambridge, England : 2003), 44(34), 15220-15231 (2015-05-15)
Anion complexation properties of two new tripodal amide receptors have been extensively studied here. Two tripodal receptors have been synthesized from the reaction of cyanobenzoyl acid chloride with two tri-amine building blocks such as (i) tris(2-aminoethyl)amine and (ii) tris(2-(4-aminophenoxy)ethyl)amine, which

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service