Skip to Content
Merck
All Photos(1)

Documents

124516

Sigma-Aldrich

Girard′s Reagent P

98%

Synonym(s):

1-(Carboxymethyl)pyridinium chloride hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10ClN3O
CAS Number:
Molecular Weight:
187.63
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

201-203 °C (dec.) (lit.)

SMILES string

[Cl-].NNC(=O)C[n+]1ccccc1

General description

Gives water-soluble derivatives of ketones and ketosteroids.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service