Skip to Content
Merck
All Photos(1)

Key Documents

11970

Sigma-Aldrich

1,3-Diphenyl-2-propenone

≥98.0% (GC)

Synonym(s):

Benzalacetophenone, Chalcone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOC6H5
CAS Number:
Molecular Weight:
208.26
Beilstein:
1210466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

solubility

dioxane: soluble 1 g/10 mL, clear, colorless

functional group

ketone
phenyl

SMILES string

O=C(\C=C\c1ccccc1)c2ccccc2

InChI

1S/C15H12O/c16-15(14-9-5-2-6-10-14)12-11-13-7-3-1-4-8-13/h1-12H/b12-11+

InChI key

DQFBYFPFKXHELB-VAWYXSNFSA-N

Gene Information

rat ... Ar(24208)

Application

1,3-Diphenyl-2-propenone was used in the preparation of pharmacologically-interesting heterocyclic systems like pyrazolines and pyrimidines[1].

Biochem/physiol Actions

1,3-Diphenyl-2-propenone (chalcone) inhibits the proliferation of human breast cancer cell lines, MCF-7 and MDA-MB-231 by inducing apoptosis and blocking cell cycle progression in the G2/M phase[2]. It is an inhibitor of Plasmodium falciparum cyclin-dependent protein kinases[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conventional and Microwave assisted Synthesis of 1, 3- Diphenyl -2- Propenone derivatives and Cytotoxic, Anti bacterial activites.
Ahmad MR and Bano N.
International Journal of ChemTech Research, 3(3), 1470-1478 (2011)
Yau-Hung Chen et al.
Molecules (Basel, Switzerland), 18(2), 2052-2060 (2013-02-07)
The aim of this study was to investigate novel chalcones with potent anti-inflammatory activities in vivo. Chalcone and two chalcone analogues (compound 5 and 9) were evaluated using a caudal fin-wounded transgenic zebrafish line "Tg(mpx:gfp)" to visualize the effect of
Lijuan Song et al.
Fitoterapia, 84, 107-114 (2012-11-20)
Hydroxysafflor yellow A (HSYA) is an active ingredient obtained from the flower of Carthamus tinctorius L. The present study investigated the effects of HSYA on lipopolysaccharide (LPS)-induced inflammatory signal transduction in human alveolar epithelial A549 cells. A549 cells stimulated with
Zijing Li et al.
Journal of medicinal chemistry, 56(2), 471-482 (2012-12-18)
Rhenium and technetium-99m cyclopentadienyl tricarbonyl complexes mimicking the chalcone structure were prepared. These complexes were proved to have affinity to β-amyloid (Aβ) in fluorescent staining on brain sections of Alzheimer's Disease (AD) patient and binding assay using Aβ(1-42) aggregates, with
Kewal Kumar et al.
Dalton transactions (Cambridge, England : 2003), 42(5), 1492-1500 (2012-10-31)
A diverse range of triazoles were prepared following well established, Cu-mediated azide-alkyne cycloaddition reactions with the aim of probing the anti-tubercular structure-activity relationships (SAR) within the β-lactam-ferrocene-triazole conjugate family. The anti-tubercular evaluation studies of the synthesized conjugates revealed that none

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service