Skip to Content
Merck
All Photos(1)

Key Documents

118265

Sigma-Aldrich

3-(p-Tolyl)propionic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€121.00

€121.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€121.00

About This Item

Linear Formula:
CH3C6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€121.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

115-118 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1ccc(CCC(O)=O)cc1

InChI

1S/C10H12O2/c1-8-2-4-9(5-3-8)6-7-10(11)12/h2-5H,6-7H2,1H3,(H,11,12)

InChI key

LDYGRLNSOKABMM-UHFFFAOYSA-N

Application

3-(p-Tolyl)propionic acid was used during the coupling of 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide with 4,5-diphenyl-2-(2-p-tolyl-ethyl)-4,5-dihydro-1H-imidazole resin[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory H Merriman et al.
Bioorganic & medicinal chemistry letters, 15(2), 435-438 (2004-12-18)
A series of 4,5-diarylimidazoline libraries were prepared using high-throughput solid-phase and microwave techniques. The compounds were evaluated as P2X(7) antagonists and their SAR is described.
Christopher S Wilson et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(50), 11593-11603 (2020-06-11)
Dinuclear metallodrugs offer much potential in the development of novel anticancer chemotherapeutics as a result of the distinct interactions possible with bio-macromolecular targets and the unique biological activity that can result. Herein, we describe the development of isostructural homo-dinuclear OsII

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service