Skip to Content
Merck
All Photos(1)

Key Documents

117641

Sigma-Aldrich

3-Aminocrotononitrile

96%

Synonym(s):

3-Amino-2-butenenitrile, 3-Iminobutyronitrile, Diacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH3)=CHCN
CAS Number:
Molecular Weight:
82.10
Beilstein:
1719815
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

solubility

95% ethanol: soluble 25 mg/mL, clear, colorless to yellow

functional group

amine
nitrile

storage temp.

2-8°C

SMILES string

C\C(N)=C/C#N

InChI

1S/C4H6N2/c1-4(6)2-3-5/h2H,6H2,1H3/b4-2+

InChI key

DELJOESCKJGFML-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Aminocrotononitrile was used as bisnucleophilic reagent which on cyclocondensation with hexafluoroacetone(ethoxycarbonylimine) forms bis(trifluoromethyl)pyrimidinones[1].

Biochem/physiol Actions

3-Aminocrotononitrile reacts with ferrocenyl-1,2-enones to form ferrocenyl pyridines[2]. It undergoes diazotization coupling reaction with p-substituted anilines to give 2-arylhydrazone-3-ketimino-butyronitriles[3].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena I Klimova et al.
Molecules (Basel, Switzerland), 17(9), 10079-10093 (2012-08-28)
The reactions of 2-cyano-3-ferrocenylacrylonitrile (1) with malononitrile (2) in a MeOH/H₂O or 2-PrOH/H₂O medium in the presence of Na₂CO₃ afforded 6-alkoxy-2-amino-4-ferrocenylpyridine-3,5-dicarbonitriles 3a,b (multi-component condensation) and 6-alkoxy-2-amino-4-ferrocenyl-3-ferrocenylmethyl-3,4-dihydropyridine-3,5-dicarbonitriles 4a,b (multi-component cyclodimerization). Analogous reactions of 1 with 2 in an MeOH/H₂O medium in
Synthesis of fluoro-containing pyrimidinones from hexafluoroacetone (ethoxycarbonylimine).
Sokolov VB and Aksinenko AY
Russian Chemical Bulletin, 54(6), 1518-1522 (2005)
Synthesis of disazo pyrazolo [1, 5-< i> a</i>] pyrimidines.
Karci F and Demircali A.
Dyes and Pigments, 74(2), 288-297 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service