Skip to Content
Merck
All Photos(1)

Key Documents

11578

Sigma-Aldrich

Boc-4,5-dehydro-Leu-OH (dicyclohexylammonium) salt

≥96.0% (HPLC)

Synonym(s):

(S)-2-(Boc-amino)-4-methyl-4-pentenoic acid (dicyclohexylammonium) salt, Boc-4,5-dehydro-L-leucine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4·C12H23N
CAS Number:
Molecular Weight:
410.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(=C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C12H23N.C11H19NO4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-7(2)6-8(9(13)14)12-10(15)16-11(3,4)5/h11-13H,1-10H2;8H,1,6H2,2-5H3,(H,12,15)(H,13,14)/t;8-/m.0/s1

InChI key

ZRIOKRZSXPPREV-WDBKTSHHSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service