All Photos(1)
About This Item
Linear Formula:
ClC6H3(NO2)CN
CAS Number:
Molecular Weight:
182.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
89-91 °C (lit.)
SMILES string
[O-][N+](=O)c1ccc(Cl)cc1C#N
InChI
1S/C7H3ClN2O2/c8-6-1-2-7(10(11)12)5(3-6)4-9/h1-3H
InChI key
HPWJUEZFOUOUEO-UHFFFAOYSA-N
Application
5-Chloro-2-nitrobenzonitrile has been used in the preparation of a series of 2,4-diamino-6-[(aralkyl and alicyclic)thio-, sulfinyl-, and sulfonyl]quinazolines by condensation reaction.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E F Elslager et al.
Journal of medicinal chemistry, 21(7), 639-643 (1978-07-01)
A series of 2,4-diamino-6-[(aralkyl and alicyclic)thio-, sulfinyl-, and sulfonyl]quinazolines was prepared via condensation of 5-chloro-2-nitrobenzonitrile or 5,6-dichloro-2-nitrobenzonitrile with the appropriate aralkyl or alicyclic thiopseudourea, reduction of the resulting 2-nitro-5-[(aralkyl or alicyclic)thio]benzonitrile with stannous chloride to the amine, and cyclization with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service