Skip to Content
Merck
All Photos(1)

Key Documents

113352

Sigma-Aldrich

1,4-Di-tert-butylbenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[C(CH3)3]2
CAS Number:
Molecular Weight:
190.32
Beilstein:
1617000
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
solid
Assay:
98%

Quality Level

Assay

98%

form

solid

bp

236 °C (lit.)

mp

76-78 °C (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)C(C)(C)C

InChI

1S/C14H22/c1-13(2,3)11-7-9-12(10-8-11)14(4,5)6/h7-10H,1-6H3

InChI key

OOWNNCMFKFBNOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-di-tert-butylbenzene (DTBB) is crystallized from several organic solvents to study the effect of solvents and crystallization conditions on its habit[1].

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N R Clement et al.
Biochemistry, 20(6), 1539-1543 (1981-03-17)
The effects of small hydrophobic molecules on valinomycin-mediated K+ transport in small unilamellar soybean phospholipid vesicles have been studied by using a vesicle-entrapped pH-sensitive hydrophilic fluorescence probe to monitor counterion-limited, passive H+ diffusion into vesicles after an abrupt decrease in
The effect of solvents on crystal habit of 1,4-DI-tert-butylbenzene (DTBB).
Garti N, et al.
Journal of Crystal Growth, 54(2), 227-231 (1981)
Christopher Elam et al.
European journal of medicinal chemistry, 46(5), 1512-1523 (2011-03-01)
Two screening protocols based on recursive partitioning and computational ligand docking methodologies, respectively, were employed for virtual screens of a compound library with 345,000 entries for novel inhibitors of the enzyme sarco/endoplasmic reticulum calcium ATPase (SERCA), a potential target for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service