About This Item
Recommended Products
vapor density
3.5 (vs air)
vapor pressure
50 mmHg ( 20 °C)
Assay
99%
form
liquid
autoignition temp.
599 °F
expl. lim.
8.5 %
availability
available only in EU
refractive index
n20/D 1.392 (lit.)
bp
84 °C (lit.)
mp
−61 °C (lit.)
solubility
water: soluble 110 g/L at 25 °C
density
0.722 g/mL at 25 °C (lit.)
SMILES string
CC(C)NC(C)C
InChI
1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3
InChI key
UAOMVDZJSHZZME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
7.8 °F - closed cup
Flash Point(C)
-13.45 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service