Skip to Content
Merck
All Photos(1)

Key Documents

109983

Sigma-Aldrich

2-Chloroethyl vinyl ether

99%, contains triethanolamine as stabilizer, contains MEHQ as stabilizer

Synonym(s):

(2-Chloroethoxy)ethene, 1-Chloro-2-vinyloxyethane, 2-Chloroethyl vinyl ether, 2-Vinyloxyethyl chloride, Vinyl 2-chloroethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€116.00

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€116.00

About This Item

Linear Formula:
ClCH2CH2OCH=CH2
CAS Number:
Molecular Weight:
106.55
Beilstein:
773787
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€116.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

760 mmHg ( 109 °C)

Assay

99%

contains

300-500 triethanolamine as stabilizer
50-100 MEHQ as stabilizer

refractive index

n20/D 1.438 (lit.)

bp

109 °C (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

ClCCOC=C

InChI

1S/C4H7ClO/c1-2-6-4-3-5/h2H,1,3-4H2

InChI key

DNJRKFKAFWSXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service