Skip to Content
Merck
All Photos(1)

Documents

10894

Sigma-Aldrich

Hexachloro-2-propanone

produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)

Synonym(s):

HCA, Hexachloroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3COCCl3
CAS Number:
Molecular Weight:
264.75
Beilstein:
1707475
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥99.0% (GC)

refractive index

n20/D 1.511 (lit.)

bp

66-70 °C/6 mmHg (lit.)

mp

−6-−2 °C (lit.)

density

1.743 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

SMILES string

ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl

InChI

1S/C3Cl6O/c4-2(5,6)1(10)3(7,8)9

InChI key

DOJXGHGHTWFZHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexachloro-2-propanone is a colorless liquid. It is present in herbicide Bromacil formulation. Hexachloro-2-propanone is precursor for the generation and cycloaddition of oxyallyl intermediates. It reverts the Ames strains TA98 and TA100 but not the non-plasmid strains TA1537, TA1535 and TA1538.

Application

Hexachloro-2-propanone (hexachloroacetone) was used in the synthesis of chirally deuterated benzyl chlorides and enol phosphate 2,2-dichloro-1-(trichloromethyl)ethenyl diethyl phosphate. It was also used in obtaining efficient amino acid activation by exploiting the rapid formation of acid chlorides under low temperature and acid/base free conditions.

Other Notes

prices for bulk quantities on request

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R Nestmann et al.
Environmental mutagenesis, 7(2), 163-170 (1985-01-01)
Reports in the literature describing artifactual results owing to interactions of test materials with solvents are becoming more frequent. The present study was initiated to examine possible interactions of 1,1,3-trichloro-, 1,1,3,3-tetrachloro-, pentachloro- and hexachloroacetones with different solvents, since certain solvent
Konstantina Sargenti et al.
Liver international : official journal of the International Association for the Study of the Liver, 35(2), 391-400 (2014-07-22)
Population-based data on the occurrence of healthcare-associated (HCA) and hospital-acquired (HA) bacterial infections in cirrhosis, their predictors, and their impact on outcome are limited. All patients with incident cirrhosis in 2001-2010 residing in an area of 600,000 inhabitants were retrospectively
Determination of hexachloroacetone in air.
E Kissa
Analytical chemistry, 55(8), 1222-1225 (1983-07-01)
Derek W Barnett et al.
Journal of labelled compounds & radiopharmaceuticals, 56(1), 6-11 (2013-11-29)
Chirally deuterated benzyl chlorides were prepared using novel, general hexachloroacetone/polymer-supported triphenylphosphine treatment of chirally deuterated benzyl alcohols. Doubly labeled protected tyrosine was obtained in 62% yield with 86% de at the α-carbon and 82% de at the β-carbon. Key in
Marian Kacerovsky et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 28(2), 134-140 (2014-03-29)
Abstract Objective: To determine the cervical fluid interleukin (IL)-6 and IL-8 levels in pregnancies complicated by preterm prelabor rupture of membranes (PPROM) and the association of these interleukins with microbial invasion of the amniotic cavity (MIAC) and histological chorioamnionitis (HCA).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service