Skip to Content
Merck
All Photos(1)

Key Documents

107239

Sigma-Aldrich

3-Methylpyridazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.514 (lit.)

bp

214 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnn1

InChI

1S/C5H6N2/c1-5-3-2-4-6-7-5/h2-4H,1H3

InChI key

MXDRPNGTQDRKQM-UHFFFAOYSA-N

General description

3-Methylpyridazine(3-Mepydz) reacts with halogenotrimethylplatinum (IV), [PtXMe3)4], to form complexes of type fac-[PtXMe3(3-Mepydz)2] (X = Cl, Br or I)[1]. It undergoes self-association in aqueous solution at acidic, neutral and basic pH [2]. It is a diazaaromatic compound and reacts with bis (1,1,1,5,5,5-hexafluoropentane-2,4-dionato) copper (II) complexes[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H H Borchert et al.
Die Pharmazie, 44(9), 625-630 (1989-09-01)
Pyridazin (1) and 3-methylpyridazine (6) undergo oxidative biotransformation in an unexpected high degree. Beside the unchanged compounds, after administration of 1 two isomeric monohydroxylated products (2, 3), 4,5-dihydrodihydroxypyridazine (4) and 4,5-dihydroxypyridazine (5) and after administration of 6 one ringhydroxylated 6-derivative
Fernando Peral et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(6), 1223-1237 (2003-03-28)
The self-association of pyridazine and pyrazine was studied in aqueous solution at acidic, neutral and basic pH values, by ultraviolet spectroscopy. The spectra of pyridazine in the mid-ultraviolet region did not show any variation in molar absorptivity upon concentration of
A dynamic NMR study of 1, 2-metallotropic shifts in trimethylplatinum (IV) halide complexes fo 3-methylpyridazine.
Abel EW, et al.
Polyhedron, 13(20), 2907-2913 (1994)
Preparation and characterisation of bis (1, 1, 1, 5, 5, 5-hexafluoropentane-2, 4-dionato) copper (II) complexes with diazaaromatic compounds. Part 1. Crystal structures and characterisation of several adducts with diazines.
Kogane T, et al.
J. Chem. Soc., Dalton Trans., 1, 13-18 (1994)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service