AABH99A42F3B
AA BLOCKS, INC.
5-Methylthieno[2,3-d]pyrimidin-4-hydrazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-hydrazino-5-methylthieno[2,3-d]pyrimidine
Inchi Code
1S/C7H8N4S/c1-4-2-12-7-5(4)6(11-8)9-3-10-7/h2-3H,8H2,1H3,(H,9,10,11)
InChI key
IWRWDRZDFILWMR-UHFFFAOYSA-N
storage temp.
2~8 C
Country of Origin
US
Product Link
https://www.aablocks.com/prod/19786-56-2
Synonym
4-HYDRAZINO-5-METHYLTHIENO[2,3-D]PYRIMIDINE
MSDS
https://www.aablocks.com/downloadSds?cas=19786-56-2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.