Merck
  • Analysis of azanaphthalenes and their enzyme oxidation products by high-performance liquid chromatography, infrared spectroscopy and mass spectrometry.
Analysis of azanaphthalenes and their enzyme oxidation products by high-performance liquid chromatography, infrared spectroscopy and mass spectrometry.
Journal of chromatography (1979-09-21)
C Stubley, J Godfrey, P Stell, D W Mathieson
PMID528640
ABSTRACT
Adsorption and reversed-phase high-performance liquid chromatography (HPLC) have been successfully used to separate metabolites from the parent heterocycles (isoquinoline, 3-methylisoquinoline, phthalazine, quinazoline, quinoxaline and cinnoline). Retention data are reported. The metabolites, hydroxyazanaphthalenes, which arise as a result of aldehyde oxidase catalysed oxidation, could be extracted in microgram quantities from incubation mixtures by n-butanol. Complete identification of the oxidation products was achieved by collecting fractions corresponding to each compound from the HPLC eluate and subjecting these to infrared and mass-spectral analysis.
Sigma-Aldrich
3-Methylisoquinoline, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
5 g
Estimated to ship on July 11, 2025
from MILWAUKEE
$35.40
20 g
Estimated to ship on July 11, 2025
from MILWAUKEE
$104.00
100 g
Estimated to ship on July 11, 2025
from MILWAUKEE
$324.00
1 kg
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$1,780.00
To order products, please contact your local dealer.