Skip to Content
Merck
All Photos(1)

Key Documents

1710006

USP

Vanillin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
Beilstein:
472792
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

API family

vanillin

manufacturer/tradename

USP

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillin is an aromatic compound, and is used for flavoring foods. It is has biogenetic relationship to phenylpropanoid pathway. It is mostly used in foods, beverages, perfumes and pharmaceuticals.

Application

It has been used as reference standard in calibrating hot stage; during thermomicroscopic investigation performed with optical polarizing microscope; usually equipped with a hot stage and temperature controller.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal architecture and physicochemical properties of felodipine solvates.
Surov, Artem O., et al.
CrystEngComm, 15.30, 6054-6061 (2013)
Biotechnological production of vanillin.
Priefert, H., J. Rabenhorst, and A. Steinbuchel
Applied Microbiology and Biotechnology, 56.3-4, 296-314 (2001)
Vanillin
Walton, Nicholas J., Melinda J. Mayer, and Arjan Narbad
Phytochemistry, 63.5, 505-515 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service