Skip to Content
Merck
All Photos(1)

Key Documents

PHL89227

Hyperoside

phyproof® Reference Substance

Synonym(s):

Quercetin 3-D-galactoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-D-galactoside, Hyperin, Hyperoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
5784795
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15+,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-DTGCRPNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Protects against peroxide-induced oxidative damage to cells by scavenging reactive oxygen species and enhancing activity of anti-oxidant enzymes, in particular, catalase and glutathione peroxidase.

Other Notes

A flavonol glycoside widely distribute in plants, including mimosa and St. John′s wort.
This compound is commonly found in plants of the genus: crataegus eucalyptus hypericum sambucus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service