Skip to Content
Merck
All Photos(1)

Key Documents

PHL89157

Aloe-emodin

phyproof® Reference Substance

Synonym(s):

1,8-Dihydroxy 3-hydroxymethylanthraquinone, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone, 3-Hydroxymethylchrysazine, Rhabarberone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
2059062
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing

format

neat

SMILES string

OCc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1

InChI

1S/C15H10O5/c16-6-7-4-9-13(11(18)5-7)15(20)12-8(14(9)19)2-1-3-10(12)17/h1-5,16-18H,6H2

InChI key

YDQWDHRMZQUTBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aloe-emodin, an anthraquinone derivative, is an active constituent of several medicinal plants. It shows gastrointestinal effects by exerting laxative activity.
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

Aloe-emodin may be used as an analytical reference standard for the determination of the analyte in plant and animal samples by high-performance liquid chromatography technique.

Biochem/physiol Actions

Laxative/cathartic compound; increases the contraction of intestinal smooth muscle by releasing endogenous acetylcholine. Anti-tumor activity is associated with an increased production of reactive oxygen species (ROS) that, in turn, reduces the mitochondrial transmembrane electrical potential, thus inducing a permeability transition that sets in motion a series of events culminating in cellular apoptosis. Genotoxicity and mutagenicity appear to be due to the inhibition of topoisomerase II activity by aloe emodin.

Other Notes

This compound is commonly found in plants of the genus: aloe

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of the anthraquinones aloe-emodin and aloin by gas chromatography/mass spectrometry
ElSohly MA, et al.
International Immunopharmacology, 4(14), 1739-1744 (2004)
Simultaneous determination and pharmacokinetic studies of aloe emodin and chrysophanol in rats after oral administration of Da-Cheng-Qi decoction by high-performance liquid chromatography
Tang WF, et al.
Biomedical Chromatography, 21(7), 701-707 (2007)
One single standard substance for the determination of multiple anthraquinone derivatives in rhubarb using high-performance liquid chromatography-diode array detection
Gao XY, et al.
Journal of Chromatography A, 1216(11), 2118-2123 (2009)
Simultaneous determination of five anthraquinones in medicinal plants and pharmaceutical preparations by HPLC with fluorescence detection
He D, et al.
Journal of Pharmaceutical and Biomedical Analysis, 49(4), 1123-1127 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service