Skip to Content
Merck
All Photos(1)

Key Documents

G005629

HPLC Analysis of Tryptic Digest of Protein Mixture on BIOshell A160 2.7 μm Peptide C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase: [A] 0.1% formic acid in water; [B] 0.4% formic acid in 25:75 (v/v) water:acetonitrile
gradient: 6 to 70% B in 40 min
flow rate: 0.3 mL/min
column temp.: 35 °C
detector: ESI(+), TOF
injection: 2 μL
sample: 10 pmol/L in 0.1% formic acid

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service