Skip to Content
Merck
All Photos(1)

Documents

97585

Supelco

Citrus Red 2

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-(2,5-Dimethoxyphenylazo)-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O3
CAS Number:
Molecular Weight:
308.33
Colour Index Number:
12156
Beilstein:
4510811
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

shipped in

wet ice

storage temp.

2-8°C

SMILES string

COc1ccc(OC)c(c1)N=Nc2c(O)ccc3ccccc23

InChI

1S/C18H16N2O3/c1-22-13-8-10-17(23-2)15(11-13)19-20-18-14-6-4-3-5-12(14)7-9-16(18)21/h3-11,21H,1-2H3

InChI key

GJUABKCEXOMRPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service