Skip to Content
Merck
All Photos(1)

Key Documents

861253

Supelco

Phenols Mix 1

in isopropanol (varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in isopropanol (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    4-Chloro-3-methylphenol 2000 μg/mL
    2-Chlorophenol 2000 μg/mL
    2,4-Dichlorophenol 2000 μg/mL
    2,6-Dichlorophenol 2000 μg/mL
    2,4-Dimethylphenol 2000 μg/mL
    2,4-Dinitrophenol 2000 μg/mL
    2-Methyl-4,6-dinitrophenol 2000 μg/mL
    o-Cresol 2000 μg/mL
    2-Nitrophenol 2000 μg/mL
    4-Nitrophenol 2000 μg/mL
    Pentachlorophenol 2000 μg/mL
    Phenol 2000 μg/mL
    2,3,4,6-Tetrachlorophenol 2000 μg/mL
    2,4,5-Trichlorophenol 2000 μg/mL
    2,4,6-Trichlorophenol 2000 μg/mL
    m-Cresol 1000 μg/mL
    p-Cresol 1000 μg/mL
    See All (17)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service