Skip to Content
Merck
All Photos(1)

Key Documents

821855

TOYOPEARL GigaCap® Q-650M Bulk Media

bottle of 250 mL, phase ammonium ion, quaternary, 75 μm particle size

Synonym(s):

TOYOPEARL GigaCap® Q-650 Anion Exchange Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
particle size:
75 μm
matrix active group:
ammonium ion, quaternary phase
matrix:
hydroxylated methacrylic polymer

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 21855

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

75 μm

operating pH

3-13

capacity

>162 g/L adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl GigaCap Q-650 resin is a high capacity, high resolution anion exchange resin* optimized for the capture and purification of both small and large proteins, including oligonucleotides and monoclonal antibodies. A Toyopearl HW-65 polymeric bead has been chemically modified to provide a higher number of anionic binding sites for increased resin capacity.
Toyopearl GigaCap Q-650M resin exhibits typical dynamic binding capacities in the vicinity of 175 mg/mL for BSA and 49 mg/mL for thyroglobulin, whereas, Toyopearl GigaCap Q-650S has dynamic binding capacities approaching 190 g/L for bovine serum albumin.
Toyopearl GigaCap Q-650 resin maintains high capacity at increased linear velocities, has good pressure-flow characteristics and will withstand back pressures up to 3 bar. The high capacity, combined with the 35 μm mean particle size, makes Toyopearl GigaCap Q-650S ideal for high resolution applications.
In addition to the high DBC and good pressure flow characteristics, Toyopearl GigaCap Q-650 resin displays excellent binding and elution kinetics, which results in a more concentrated protein fraction collected during the elution step.
*specifically designed for packed bed use

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl GigaCap is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service