Skip to Content
Merck
All Photos(1)

Key Documents

819823

TOYOPEARL® SuperQ-650S Bulk Media

phase ammonium ion, quaternary, bottle of 25 mL, 35 μm particle size

Synonym(s):

TOYOPEARL® SuperQ-650 Anion Exchange Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
particle size:
35 μm
matrix active group:
ammonium ion, quaternary phase
matrix:
hydroxylated methacrylic polymer

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 19823

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

35 μm

operating pH

2-13

capacity

130 g/L±25 adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SuperQ-650 resin is a strong anion exchanger for biomolecule purification. It can be used for high throughput capture, interresinte purification, and polishing process steps. If more powerful resolution is needed the smaller particle TSKgel SuperQ-5PW resin should be used. Selectivity remains the same.

Specifications

Clean in place with 0.1 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service