Skip to Content
Merck
All Photos(1)

Key Documents

48270-U

Supelco

HPLC Isocratic Systems Diagnostics Mix

certified reference material, in methanol: water (3:2) (varied conc.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

concentration

in methanol: water (3:2) (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

for use with HPLC System Diagnostics

format

multi-component solution

storage temp.

2-30°C

General description

This formulation is designed for evaluating how reliably a chromatographic system is providing such fundamentally important parameters as flow rate, proportioning, and mixing.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Butylparaben 25 μg/mL
    Ethyl 4-hydroxybenzoate 14 μg/mL
    Methyl 4-hydroxybenzoate 10 μg/mL
    Propyl 4-hydroxybenzoate 15 μg/mL

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

9.99 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service