Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1143

Sigma-Aldrich

Anti-RPE65 Antibody, clone 401.8B11.3D9 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

All-trans-retinyl-palmitate hydrolase, EC:3.1.1.64, Lutein isomerase, Meso-zeaxanthin isomerase, Retinal pigment epithelium-specific 65 kDa protein, Retinoid isomerohydrolase, Retinol isomerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

401.8B11.3D9, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 60.94 kDa
observed mol wt ~60 kDa

purified by

using protein G

species reactivity

human, bovine

species reactivity (predicted by homology)

rat, Xenopus, canine, mouse, porcine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 401.8B11.3D9 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects RPE65.

Immunogen

Bovine RPE65 from microsomal membrane preparations.

Application

Quality Control Testing

Evaluated by Western Blotting in Bovine retina tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RPE65 in Bovine retina tissue lysate.

Tested Applications

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected RPE65 in Human retina tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected RPE65 in Human retina tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Retinoid isomerohydrolase (UniProt: Q28175; also known as EC:3.1.1.64, All-trans-retinyl-palmitate hydrolase, Lutein isomerase, Meso-zeaxanthin isomerase, Retinal pigment epithelium-specific 65 kDa protein, Retinol isomerase) is encoded by the RPE65 gene (Gene ID: 282043) in bovine. Retinol isomerase is a lipid-anchor protein that is attached to the membrane when palmitoylated and is present in soluble form when unpalmitoylated.  It is shown to be critical for retinoid cycle and is involved in the regeneration of 11-cis-retinal, the chromophore of rod and cone opsins. It catalyzes the cleavage and isomerization of all-trans-retinyl fatty acid esters to 11-cis-retinol that is further oxidized by 11-cis retinol dehydrogenase to 11-cis-retinal for use as visual chromophore. It can also catalyze the isomerization of lutein to meso-zeaxanthin an eye-specific carotenoid. The soluble form is reported to binds vitamin A (all-trans-retinol), making it available for lecithin retinol acyltransferase (LRAT) processing to all-trans-retinyl ester. The membrane form, palmitoylated by LRAT, binds all-trans-retinyl esters, making them available for isomerohydrolase processing to all-cis-retinol. The soluble form can be regenerated by transferring its palmitoyl groups onto 11-cis-retinol, a reaction catalyzed by LRAT. It undergoes light-dependent intracellular transport to become more concentrated in the central region of the retina pigment epithelium cells. Mutations in human RPE65 gene are known to cause Retinitis pigmentosa that is characterized by retinal pigment deposits visible on fundus examination and primary loss of rod photoreceptor cells followed by secondary loss of cone photoreceptors.  This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.:  Golczak, M., et al. (2010). J. Biol. Chem. 285(13); 9667-9682; Jin, M., et al. (2005). Cell. 122(3); 449-459).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service