Key Documents
Z744002
Worthington Accessories
rack for 8 drawers for LS4800 liquid nitrogen refrigerator
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Worthington R05K-9C50
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Could you please send me the dimensions of this rack and the dewar in which the rack will fit? How does the rack hang in the dewar? Thank you.
    1 answer
    1. Dimensions: This rack has an overall length of 30.41 inches. This rack handle is 12.91 inches and the rack is 18.5 inches. The area (L x W) of the rack is 2.69" x 2.69”. There is a ring in the upper portion of the Dewar that the handle hangs from. This rack is suitable for product Z743997 which is 35 inches in height.
      Helpful?